Proteintech
Manufacturer Code:141741AP150UL
Catalog # 50-172-9264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | XPR1 Fusion Protein Ag5373 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein SYG1 homolog; Protein SYG1 homolog SYG1 X receptor X3 XPR1 XR; receptor for MCF virus 1; Rmc-1; suppressor of yeast G<a> deletion; susceptibility to xenotropic virus; X-receptor; xenotropic and polytropic murine leukemia virus receptor; Xenotropic and polytropic murine leukemia virus receptor X3; Xenotropic and polytropic retrovirus receptor 1; xenotropic and polytropic retrovirus receptor 1 homolog
Gene Aliases: IBGC6; Rmc-1; Rmc1; Sxv; SYG1; X3; XPR1; XR
UniProt ID: (Human) Q9UBH6, (Mouse) Q9Z0U0
Entrez Gene ID: (Human) 9213, (Mouse) 19775, (Rat) 289424
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.