Invitrogen
Manufacturer Code:PA514639
Catalog # PIPA514639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 137-166 amino acids from the N-terminal region of human XPOT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exportin(tRNA); exportin, tRNA (nuclear export receptor for tRNAs); Exportin-T; Exportin-T Exportin-T tRNA exportin Exportin(tRNA) XPOT; tRNA exportin
Gene Aliases: XPO3; XPOT
UniProt ID: (Human) O43592
Entrez Gene ID: (Human) 11260
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.