Abnova
Manufacturer Code:H00057510M01A
Catalog # 89-107-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C5-1B3 |
Immunogen | XPO5 (AAH001291 a.a. &bsim 280 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant XPO5.
For Research Use Only
Protein Aliases: Exp5; Exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; Ran-binding protein 21
Gene Aliases: exp5; KIAA1291; RANBP21; XPO5
UniProt ID: (Human) Q9HAV4
Entrez Gene ID: (Human) 57510
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.