Proteintech
Manufacturer Code:156551AP150UL
Catalog # 50-173-0380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | XPNPEP3 Fusion Protein Ag8007 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aminopeptidase P3; Aminopeptidase P3 APP3 X Pro aminopeptidase 3 XPNPEP3; APP3; Intermediate Cleaving Peptidase 55; probable Xaa-Pro aminopeptidase 3; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3, mitochondrial; Xaa-Pro aminopeptidase 3
Gene Aliases: APP3; E430012M05Rik; ICP55; NPHPL1; XPNPEP3
UniProt ID: (Human) Q9NQH7, (Rat) B5DEQ3, (Mouse) B7ZMP1
Entrez Gene ID: (Human) 63929, (Rat) 685823, (Mouse) 321003
Molecular Function: hydrolase metalloprotease nucleic acid binding protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.