Invitrogen
Manufacturer Code:PA583142
Catalog # PIPA583142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human XPNPEP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; Cytosolic aminopeptidase P; sAmp; Soluble aminopeptidase P; X-Pro aminopeptidase 1; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1, soluble; Xaa-Pro aminopeptidase 1; XPNPEP XPNPEPL XPNPEPL1
Gene Aliases: APP1; SAMP; XPNPEP; XPNPEP1; XPNPEPL; XPNPEPL1
UniProt ID: (Human) Q9NQW7
Entrez Gene ID: (Human) 7511
Molecular Function: hydrolase metalloprotease nucleic acid binding protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.