Invitrogen
Manufacturer Code:PA519139
Catalog # PIPA519139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (LIRETQPISKQH) corresponding to the C-terminus amino acids of XPNPEP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; cytoplasmic aminopeptidase P; Cytosolic aminopeptidase P; sAmp; Soluble aminopeptidase P; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1, soluble; Xaa-Pro aminopeptidase 1; xpnpep1; XPNPEP1 X-prolyl aminopeptidase (aminopeptidase P) 1 soluble SAMP XPNPEP XPNPEPL XPNPEPL1 OTTHUMP00000020457 OTTHUMP00000058856 X-prolyl aminopeptidase (aminopeptidase P) 1 soluble (SAMP XPNPEP XPNPEPL) X-prolyl aminopeptidase (aminopeptidase P)-like; xpnpep1 {ECO:0000312|RGD:621274}
Gene Aliases: App; APP1; D230045I08Rik; SAMP; XPNPEP; XPNPEP1; XPNPEPL; XPNPEPL1
UniProt ID: (Human) Q9NQW7, (Mouse) Q6P1B1, (Rat) O54975
Entrez Gene ID: (Human) 7511, (Pig) 100152688, (Mouse) 170750, (Rat) 170751
Molecular Function: hydrolase metalloprotease nucleic acid binding protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.