Invitrogen
Manufacturer Code:PA5110585
Catalog # PIPA5110585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 44-240 of human XPNPEP1 (NP_065116.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aminoacylproline aminopeptidase; Aminoacylproline aminopeptidase App APP1 Cytosolic aminopeptidase P D230045I08Rik OTTHUMP00000020457 OTTHUMP00000058856 sAmp Soluble aminopeptidase P Xaa-Pro aminopeptidase 1 XPNPEP Xpnpep1 XPNPEPL XPNPEPL1 X-Pro aminopeptidase 1 X-prolyl aminopeptidase 1 soluble; aminopeptidase P, cytosolic; Cytosolic aminopeptidase P; sAmp; Soluble aminopeptidase P; X-Pro aminopeptidase 1; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1, soluble; Xaa-Pro aminopeptidase 1; xpnpep1
Gene Aliases: APP1; D230045I08Rik; SAMP; XPNPEP; XPNPEP1; XPNPEPL; XPNPEPL1
UniProt ID: (Human) Q9NQW7, (Mouse) Q6P1B1
Entrez Gene ID: (Human) 7511, (Mouse) 170750
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.