Proteintech
Manufacturer Code:106611AP150UL
Catalog # 50-172-6489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | XPNPEP1 Fusion Protein Ag1015 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; cytoplasmic aminopeptidase P; Cytosolic aminopeptidase P; Cytosolic aminopeptidase P SAMP Soluble aminopeptidase P X Pro aminopeptidase 1 Xaa Pro aminopeptidase 1 XPNPEP XPNPEP1 XPNPEPL XPNPEPL1 XPP1; sAmp; Soluble aminopeptidase P; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1, soluble; Xaa-Pro aminopeptidase 1; xpnpep1; xpnpep1 {ECO:0000312|RGD:621274}
Gene Aliases: App; APP1; D230045I08Rik; SAMP; XPNPEP; XPNPEP1; XPNPEPL; XPNPEPL1
UniProt ID: (Human) Q9NQW7, (Mouse) Q6P1B1, (Rat) O54975
Entrez Gene ID: (Human) 7511, (Mouse) 170750, (Rat) 170751
Molecular Function: hydrolase metalloprotease nucleic acid binding protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.