OriGene
Manufacturer Code:CF503262
Catalog # 50-166-2007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B2 |
Immunogen | Full length human recombit protein of human ERCC4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA excision repair protein ERCC-4; DNA repair endonuclease XPF; DNA repair protein complementing XP-F cells; ERCC11 FANCQ RAD1 XFEPS XPF; excision repair cross-complementation group 4; excision repair cross-complementing rodent repair deficiency, complementation group 4; excision-repair, complementing defective, in Chinese hamster; Xeroderma pigmentosum group F-complementing protein; xeroderma pigmentosum, complementation group F
Gene Aliases: ERCC11; ERCC4; FANCQ; RAD1; XFEPS; XPF
UniProt ID: (Human) Q92889
Entrez Gene ID: (Human) 2072
Molecular Function: DNA binding protein endodeoxyribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.