Invitrogen
Manufacturer Code:MA537877
Catalog # PIMA537877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2401 |
Immunogen | Recombinant protein of human XPD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AA407812 AU020867 AW240756 basic transcription factor 2 80 kDa subunit BTF2 p80 COFS2 CXPD DNA excision repair protein ERCC-2 DNA repair protein complementing XP-D cells EM9 ERCC excision repair 2 TFIIH core complex helicase subunit Ercc2 Ercc-2 excision repair cross-complementation group 2 excision repair cross-complementing rodent repair deficiency complementation group 2 General transcription and DNA repair factor IIH helicase subunit XPD MGC102762 MGC126218 MGC126219 TFIIH TFIIH 80 kDa subunit TFIIH basal transcription factor complex 80 kDa subunit TFIIH basal transcription factor complex helicase subunit TFIIH basal transcription factor complex helicase XPB subunit TFIIH basal transcription factor complex helicase XPD subunit TFIIH p80 TFIIH subunit XPD TTD TTD1 xeroderma pigmentosum complementary group D xeroderma pigmentosum group D-complementing protein XPD XPDC; Basic transcription factor 2 80 kDa subunit; BTF2 p80; CXPD; DNA excision repair protein ERCC-2; DNA repair protein complementing XP-D cells; excision repair cross-complementation group 2; excision repair cross-complementing rodent repair deficiency, complementation group 2; General transcription and DNA repair factor IIH helicase subunit XPD; TFIIH 80 kDa subunit; TFIIH basal transcription factor complex 80 kDa subunit; TFIIH basal transcription factor complex helicase subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH basal transcription factor complex helicase XPD subunit; TFIIH p80; TFIIH subunit XPD; xeroderma pigmentosum complementary group D; Xeroderma pigmentosum group D-complementing protein
Gene Aliases: AA407812; AU020867; AW240756; COFS2; CXPD; EM9; Ercc-2; ERCC2; TFIIH; TTD; TTD1; XPD; XPDC
UniProt ID: (Human) P18074, (Mouse) O08811
Entrez Gene ID: (Human) 2068, (Mouse) 13871, (Rat) 308415
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.