Novus Biologicals
Manufacturer Code:NBP274876FR
Catalog # NBP274876FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D6 |
Immunogen | Full length recombinant protein of human XLF (NP_079058) produced in HEK293T cells. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cernunnos non-homologous end-joining factor 1 nonhomologous end-joining factor 1 Protein cernunnos XLFFLJ12610 XRCC4-like factor; Non-homologous end-joining factor 1; nonhomologous end-joining factor 1; Protein cernunnos; XRCC4-like factor
Gene Aliases: NHEJ1; XLF
UniProt ID: (Human) Q9H9Q4
Entrez Gene ID: (Human) 79840
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.