Proteintech
Manufacturer Code:270491AP150UL
Catalog # 50-173-5541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | XKRX Fusion Protein Ag25266 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Membrane protein XPLAC; Membrane protein XPLAC XK related protein 2 XKR2 XKRX XPLAC XRG2; X Kell blood group precursor-related, X-linked; X Kell blood group-related, X-linked; X-linked Kx blood group related, X-linked; XK, Kell blood group complex subunit-related, X-linked; XK-related protein 2
Gene Aliases: XKR2; XKRX; XPLAC; XRG2
UniProt ID: (Human) Q6PP77
Entrez Gene ID: (Human) 402415
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.