Invitrogen
Manufacturer Code:PA590227
Catalog # PIPA590227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 59-138 of human XK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kell blood group precursor (McLeod phenotype) homolog; McLeod syndrome gene homolog; Membrane transport protein XK; X-linked Kx blood group (McLeod syndrome) homolog; XK homolog; XK-related protein 1
Gene Aliases: 1810038K19Rik; Xk; Xkh; Xkr1; XRG1
UniProt ID: (Mouse) Q9QXY7, (Rat) Q5GH61
Entrez Gene ID: (Mouse) 22439, (Rat) 497078
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.