Proteintech
Manufacturer Code:128301AP150UL
Catalog # 50-172-8283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | XK Fusion Protein Ag3550 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kell blood group precursor (McLeod phenotype); Kell blood group precursor (McLeod phenotype) homolog; Kell complex 37 kDa component; Kell complex 37 kDa component KX Kx antigen Membrane transport protein XK X1k XK XK related protein 1 XKR1 XRG1; Kx antigen; McLeod syndrome gene homolog; Membrane transport protein XK; neuroacanthocytosis; neurocanthocytosis; X-linked Kx blood group (McLeod syndrome) homolog; XK homolog; XK, Kell blood group complex subunit (McLeod syndrome); XK-related protein 1
Gene Aliases: 1810038K19Rik; KX; MCLDS; NA; NAC; X1k; XK; Xkh; XKR1; XRG1
UniProt ID: (Human) P51811, (Mouse) Q9QXY7, (Rat) Q5GH61
Entrez Gene ID: (Human) 7504, (Mouse) 22439, (Rat) 497078
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.