Invitrogen
Manufacturer Code:PA557072
Catalog # PIPA557072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human XIRP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-xin; cardiomyopathy associated 3; Cardiomyopathy-associated protein 3; myomaxin; Xeplin; xin actin-binding repeat containing 2; Xin actin-binding repeat-containing protein 2; xin repeat protein 2
Gene Aliases: CMYA3; XIRP2
UniProt ID: (Human) A4UGR9
Entrez Gene ID: (Human) 129446
Molecular Function: actin family cytoskeletal protein cytoskeletal protein nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.