Invitrogen
Manufacturer Code:PA553805
Catalog # PIPA553805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human XIRP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiomyopathy associated 1; Cardiomyopathy-associated protein 1; xin actin-binding repeat containing 1; Xin actin-binding repeat-containing protein 1; Xin actin-binding repeat-containing protein 1 Cardiomyopathy-associated protein 1 XIRP1 CMYA1 XIN
Gene Aliases: CMYA1; XIN; XIRP1
UniProt ID: (Human) Q702N8
Entrez Gene ID: (Human) 165904
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.