Invitrogen
Manufacturer Code:PA522812
Catalog # PIPA522812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a portion of human ILP-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Baculoviral IAP repeat-containing protein 4; E3 ubiquitin-protein ligase XIAP; IAP-3; IAP-like protein; ILP; Inhibitor of apoptosis protein 3; RING-type E3 ubiquitin transferase XIAP; RP1-315G1.5 API3 BIRC4 IAP-3 ILP1 MIHA XLP2 hIAP-3 hIAP3 E3 ubiquitin-protein ligase XIAP IAP-like protein X-linked IAP X-linked inhibitor of apoptosis protein baculoviral IAP repeat-containing 4 baculoviral IAP repeat-containing protein 4 inhibitor of apoptosis protein 3; X-linked IAP; X-linked inhibitor of apoptosis protein; X-linked inhibitor of apoptosis, E3 ubiquitin protein ligase
Gene Aliases: API3; BIRC4; hIAP-3; hIAP3; IAP-3; IAP3; ILP1; MIHA; XIAP; XLP2
UniProt ID: (Human) P98170
Entrez Gene ID: (Human) 331
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.