Novus Biologicals
Manufacturer Code:NB10056183SS
Catalog # NB10056183T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant BIR2 domain protein fragment of human XIAP was used as immunogen. The BIR2 domain used for immunogen corresponds to amino acids 163-230 of human XIAP (Deveraux et al 1999). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: API3X-linked inhibitor of apoptosis protein apoptosis inhibitor 3 baculoviral IAP repeat-containing 4 baculoviral IAP repeat-containing protein 4 BIRC4XLP2 E3 ubiquitin-protein ligase XIAP EC 6.3.2.- hIAP3 hIAP-3 hILP IAP3 IAP-3 IAP-like protein ILP ILP1 Inhibitor of apoptosis protein 3 MIHA X-linked IAP X-linked inhibitor of apoptosis; Baculoviral IAP repeat-containing protein 4; E3 ubiquitin-protein ligase XIAP; IAP-3; IAP-like protein; ILP; Inhibitor of apoptosis protein 3; RING-type E3 ubiquitin transferase XIAP; X-linked IAP; X-linked inhibitor of apoptosis protein; X-linked inhibitor of apoptosis, E3 ubiquitin protein ligase
Gene Aliases: API3; BIRC4; hIAP-3; hIAP3; IAP-3; IAP3; ILP1; MIHA; XIAP; XLP2
UniProt ID: (Human) P98170
Entrez Gene ID: (Human) 331
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.