Abnova
Manufacturer Code:PAB28089
Catalog # 89-129-594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified XDH from bovine buttermilk Xanthine. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against native XDH (Bovine).
For Research Use Only
Protein Aliases: Xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; Xanthine oxidase; Xanthine oxidoreductase; XD; XO; XOR
Gene Aliases: XDH; XOR
UniProt ID: (Bovine) P80457
Entrez Gene ID: (Bovine) 280960
Molecular Function: dehydrogenase oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.