Invitrogen
Manufacturer Code:MA534780
Catalog # PIMA534780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG38-40 |
Immunogen | Recombinant protein within human Xanthine Oxidase aa 100-300. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: BOS_11417 Xanthine dehydrogenase Xanthine dehydrogenase/oxidase Xanthine oxidase Xanthine oxidoreductase XD XDH xdh antibody XDHA xdha antibody XO XOR xor antibody Xox1 Xox-1; Xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; Xanthine oxidase; Xanthine oxidoreductase; XD; XO; XOR
Gene Aliases: XDH; XDHA; XO; XOR
UniProt ID: (Human) P47989
Entrez Gene ID: (Human) 7498
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.