Invitrogen
Manufacturer Code:PA570621
Catalog # PIPA570621
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat XDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Xanthine dehydrogenase; Xanthine dehydrogenase antibody Xanthine oxidase antibody Xanthine oxidoreductase antibody xdh antibody xdha antibody xor antibody; Xanthine dehydrogenase/oxidase; Xanthine oxidase; Xanthine oxidoreductase; XD; XO; XOR
Gene Aliases: XDH; XDHA; XO; XOR; Xox-1; Xox1
UniProt ID: (Bovine) P80457, (Human) P47989, (Rat) P22985, (Mouse) Q00519
Entrez Gene ID: (Dog) 483028, (Bovine) 280960, (Rabbit) 100144327, (Goat) 100861280, (Human) 7498, (Rat) 497811, (Guinea pig) 100731391, (Mouse) 22436
Molecular Function: dehydrogenase oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.