Abnova
Manufacturer Code:PAB16226
Catalog # 89-118-112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human XCR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of XCR1.
For Research Use Only
Protein Aliases: CCXCR1; chemokine (C motif) receptor 1; chemokine (C motif) XC receptor 1; Chemokine XC receptor 1; G protein-coupled receptor 5; G-protein coupled receptor 5; GPR5; Lymphotactin receptor; XC chemokine receptor 1
Gene Aliases: CCXCR1; GPR5; XCR1
UniProt ID: (Human) P46094
Entrez Gene ID: (Human) 2829
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.