Novus Biologicals
Manufacturer Code:NBP212004C
Catalog # NBP212004C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0604-4G24 |
Immunogen | Human Lymphotactin |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ATAC; ATACSmall-inducible cytokine C1 C motif chemokine 1 chemokine (C motif) ligand 1 Cytokine SCM-1 LPTNSCM-1-alpha LTNXC chemokine ligand 1 lymphotactin lymphotaxin SCM-1 SCM-1a SCM1A SCYC1SCM1 single cysteine motif 1a small inducible cytokine subfamily C member 1 (lymphotactin); C motif chemokine 1; chemokine (C motif) ligand 1; Cytokine SCM-1; Lymphotactin; Lymphotaxin; SCM-1-alpha; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); Small-inducible cytokine C1; XC chemokine ligand 1
Gene Aliases: ATAC; LPTN; LTN; SCM-1; SCM-1a; SCM1; SCM1A; SCYC1; XCL1
UniProt ID: (Human) P47992
Entrez Gene ID: (Human) 6375
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.