Abnova
Manufacturer Code:PAB19528
Catalog # 89-121-056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at C-terminus of human XCL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 200&muL of deionized water to a concentration of 500&mug/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of XCL1.
For Research Use Only
Protein Aliases: ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; Cytokine SCM-1; LPTN; LTN; Lymphotactin; Lymphotaxin; SCM-1; SCM-1-alpha; SCM-1a; SCM1; SCYC1; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); Small-inducible cytokine C1; XC chemokine ligand 1
Gene Aliases: ATAC; LPTN; LTN; SCM-1; SCM-1a; SCM1; SCM1A; SCYC1; XCL1
UniProt ID: (Human) P47992
Entrez Gene ID: (Human) 6375
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.