R&D Systems
Manufacturer Code:MAB4257SP
Catalog # MAB4257SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 525904 |
Immunogen | <EM>E. coli</EM>-derived recombinant human XBP1S Ser188-Leu287 Accession # NP_001073007 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HTF TREB5 X-box binding protein 1 X-box-binding protein 1 XBP-1 XBP2 XBP2Tax-responsive element-binding protein 5; Tax-responsive element-binding protein 5; TREB-5; X-box-binding protein 1; X-box-binding protein 1, cytoplasmic form; X-box-binding protein 1, luminal form; XBP-1
Gene Aliases: TREB-5; TREB5; XBP-1; XBP1; XBP2
UniProt ID: (Human) P17861
Entrez Gene ID: (Human) 7494
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.