Invitrogen
Manufacturer Code:PA518940
Catalog # PIPA518940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NHSWEDTFANELFPQ) corresponding to the internal amino acids of XBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Tax-responsive element-binding protein 5; TREB-5; X-box-binding protein 1; X-box-binding protein 1, cytoplasmic form; X-box-binding protein 1, luminal form; XBP-1; XBP1 TREB5 X-box binding protein 1 XBP2 OTTHUMP00000028682 X-box-binding protein-1
Gene Aliases: TREB-5; TREB5; XBP-1; XBP1; XBP2
UniProt ID: (Human) P17861
Entrez Gene ID: (Human) 7494
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.