Proteintech
Manufacturer Code:259971AP150UL
Catalog # 50-173-5156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | XBP1 Fusion Protein Ag21714 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hepatocarcinogenesis-related transcription factor; HTF; Tax-responsive element-binding protein 5; tax-responsive element-binding protein 5 homolog; TREB-5; TREB5 X box binding protein 1 XBP 1 XBP1 XBP2; X-box-binding protein 1; X-box-binding protein 1, cytoplasmic form; X-box-binding protein 1, luminal form; XBP-1
Gene Aliases: D11Ertd39e; HTF; TREB-5; TREB5; XBP-1; XBP1; XBP2
UniProt ID: (Human) P17861, (Rat) Q9R1S4, (Mouse) O35426
Entrez Gene ID: (Human) 7494, (Rat) 289754, (Mouse) 22433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.