Invitrogen
Manufacturer Code:PA5100368
Catalog # PIPA5100368
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human XAB2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610041O14Rik Adapter protein ATH-55 Ath55 AV025587 crn-related protein kim1 DKFZp762C1015 HCNP HCNP protein HCRN KIAA1177 NTC90 PP3898 Pre-mRNA-splicing factor SYF1 Protein HCNP Syf1 SYF1 homolog RNA splicing factor SYF1 pre-mRNA-splicing factor Xab2 XPA binding protein 2 XPA-binding protein 2; crn-related protein kim1; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; XPA-binding protein 2
Gene Aliases: 0610041O14Rik; AV025587; HCNP; HCRN; KIAA1177; NTC90; PP3898; SYF1; XAB2
UniProt ID: (Human) Q9HCS7, (Mouse) Q9DCD2
Entrez Gene ID: (Human) 56949, (Mouse) 67439
Molecular Function:
DNA binding protein
RNA binding protein
damaged DNA-binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.