Novus Biologicals
Manufacturer Code:NB100794
Catalog # NB100794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SVPAAVFGSLKED corresponding to C-Terminus according to NP_064581.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: crn-related protein kim1; DKFZp762C1015 HCNPcrn-related protein kim1 HCRN KIAA1177 NTC90 Protein HCNP SYF1 homolog RNA splicing factor SYF1pre-mRNA-splicing factor SYF1 XPA binding protein 2 XPA-binding protein 2; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; XPA-binding protein 2
Gene Aliases: HCNP; HCRN; KIAA1177; NTC90; PP3898; SYF1; XAB2
UniProt ID: (Human) Q9HCS7
Entrez Gene ID: (Human) 56949
Molecular Function: DNA binding protein RNA binding protein damaged DNA-binding protein mRNA processing factor mRNA splicing factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.