Abnova
Manufacturer Code:PAB6025
Catalog # 89-132-072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human XAB2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of XAB2.
For Research Use Only
Protein Aliases: crn-related protein kim1; DKFZp762C1015; HCNP; HCRN; NTC90; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; XPA-binding protein 2
Gene Aliases: HCNP; HCRN; KIAA1177; NTC90; PP3898; SYF1; XAB2
UniProt ID: (Human) Q9HCS7
Entrez Gene ID: (Human) 56949
Molecular Function:
DNA binding protein
RNA binding protein
damaged DNA-binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.