Invitrogen
Manufacturer Code:PA547658
Catalog # PIPA547658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse Wnt-4 Ile37-Glu76 Arg222-Thr295 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-4; signal molecule; wingless-related MMTV integration site 4; WNT-4 Protein Wnt-4 SERKAL
Gene Aliases: Wnt-4; Wnt4
UniProt ID: (Mouse) P22724
Entrez Gene ID: (Mouse) 22417
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.