Invitrogen
Manufacturer Code:PA537320
Catalog # PIPA537320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human Wnt3a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC119418 MGC119419 MGC119420 Protein Wnt 3a Precursor Protein Wnt-3a Wingless type MMTV integration site family member 3A Wnt 3a wnt3a WNT3A protein WNT3A; Protein Wnt-3a; vestigial tail; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3A
Gene Aliases: vt; Wnt-3a; WNT3A
UniProt ID: (Human) P56704, (Mouse) P27467
Entrez Gene ID: (Human) 89780, (Mouse) 22416
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.