Invitrogen
Manufacturer Code:PA547362
Catalog # PIPA547362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse Wnt-11 peptide Leu39-Ala79 and Lys225-Arg297 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; WNT11 HWNT11 Protein Wnt-11
Gene Aliases: HWNT11; Wnt-11; WNT11
UniProt ID: (Human) O96014, (Mouse) P48615
Entrez Gene ID: (Human) 7481, (Mouse) 22411
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.