R&D Systems
Manufacturer Code:MAB3669
Catalog # MAB3669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 406612 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Wnt-9b Ala46-Glu90 & Lys228-Tyr298 Accession # O35468.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-14b; Protein Wnt-14b wingless-type MMTV integration site family member 15 wingless-type MMTV integration site family member 9B Wnt-14b WNT14Bprotein Wnt-9b Wnt-15 WNT15Protein Wnt-15 Wnt9b; Protein Wnt-15; Protein Wnt-9b; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15
Gene Aliases: UNQ6973/PRO21956; WNT14B; WNT15; WNT9B
UniProt ID: (Human) O14905
Entrez Gene ID: (Human) 7484
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.