R&D Systems
Manufacturer Code:MAB3157
Catalog # MAB3157
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 326711 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Wnt-9a Pro48-Glu92 & Arg231-Phe304 Accession # O14904.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC138165 MGC141991 Protein Wnt-14 wingless-type MMTV integration site family member 14 wingless-type MMTV integration site family member 9A Wnt-14 WNT14protein Wnt-9a Wnt9a; Protein Wnt-14; Protein Wnt-9a; wingless-type MMTV integration site family member 9A; wingless-type MMTV integration site family, member 14; wingless-type MMTV integration site family, member 9A
Gene Aliases: WNT14; WNT9A
UniProt ID: (Human) O14904
Entrez Gene ID: (Human) 7483
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.