R&D Systems
Manufacturer Code:AF2248SP
Catalog # AF2248SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Wnt-8a peptide Trp23-Glu53 Leu196-Arg273 Accession # Q64527 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-8a; protein Wnt-8a Protein Wnt-8d wingless-type MMTV integration site family member 8A Wnt8a WNT8DWNT8d; Protein Wnt-8d; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; WNT8d
Gene Aliases: WNT8A; WNT8D
UniProt ID: (Human) Q9H1J5
Entrez Gene ID: (Human) 7478
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.