Novus Biologicals
Manufacturer Code:NBP212001JF646
Catalog # NBP22001646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0359-21D12 |
Immunogen | Mouse Wnt4 |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: Protein Wnt-4; protein Wnt-4 SERKAL wingless-type MMTV integration site family member 4 WNT-4; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4
Gene Aliases: SERKAL; UNQ426/PRO864; WNT-4; WNT4
UniProt ID: (Human) P56705
Entrez Gene ID: (Human) 54361
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.