R&D Systems
Manufacturer Code:MAB4751SP
Catalog # MAB4751SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 55025 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Wnt-4 Ile37-Glu76 Accession # NP_033549 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-4; protein Wnt-4 SERKAL wingless-type MMTV integration site family member 4 Wnt4 WNT-4; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4
Gene Aliases: SERKAL; UNQ426/PRO864; WNT-4; WNT4
UniProt ID: (Human) P56705
Entrez Gene ID: (Human) 54361
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.