R&D Systems
Manufacturer Code:IC13242G100UG
Catalog # IC13242G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 217804.2R |
Immunogen | <em>E.coli</em>-derived recombinant mouse Wnt-3a Ser36-Gln75 Trp219-Arg269 Accession # P27467 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: MGC119418 MGC119419 MGC119420 protein Wnt-3a wingless-type MMTV integration site family member 3A Wnt3a; Protein Wnt-3a; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3A
Gene Aliases: WNT3A
UniProt ID: (Human) P56704
Entrez Gene ID: (Human) 89780
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.