R&D Systems
Manufacturer Code:MAB71961SP
Catalog # MAB71961SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 793116 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Wnt-10b Asn29-Lys389 Accession # O00744 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-10b; protein Wnt-10b Protein Wnt-12 SHFM6WNT-10B protein wingless-type MMTV integration site family member 10B Wnt10b Wnt-12 WNT12; Protein Wnt-12; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; WNT-10B protein
Gene Aliases: SHFM6; WNT-12; WNT10B; WNT12
UniProt ID: (Human) O00744
Entrez Gene ID: (Human) 7480
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.