Novus Biologicals
Manufacturer Code:NBP169116
Catalog # NBP169116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to Wnt10a (wingless related MMTV integration site 10a) The peptide sequence was selected from the middle region of Wnt10a. Peptide sequence RDQRWNCSSLETRNKVPYESPIFSRGFRESAFAYAIAAAGVVHAVSNACA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ14301 protein Wnt-10a SSPS wingless-type MMTV integration site family member 10A; Protein Wnt-10a; wingless-type MMTV integration site family member 10A; wingless-type MMTV integration site family, member 10A
Gene Aliases: OODD; SSPS; STHAG4; WNT10A
UniProt ID: (Human) Q9GZT5
Entrez Gene ID: (Human) 80326
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.