Invitrogen
Manufacturer Code:PA537972
Catalog # PIPA537972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RQRLHKEYPEGDSKN from the internal region near the N-terminus of mouse Wfdc1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Prostate stromal protein ps20; ps20 growth inhibitor; WAP four-disulfide core domain 1 ps20 WAP four-disulfide core domain 1 homolog WAP four-disulfide core domain protein 1 prostate stromal protein ps20 ps20 growth inhibitor 2310058A03Rik; WAP four-disulfide core domain protein 1
Gene Aliases: 2310058A03Rik; Ps20; Wfdc1
UniProt ID: (Mouse) Q9ESH5
Entrez Gene ID: (Mouse) 67866
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.