Novus Biologicals
Manufacturer Code:NBP15308920UL
Catalog # NBP15308920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to WWP2(WW domain containing E3 ubiquitin protein ligase 2) The peptide sequence was selected from the middle region of WWP2. Peptide sequence SSASTDHDPLGPLPPGWEKRQDNGRVYYVNHNTRTTQWEDPRTQGMIQEP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIP2; AIP2Nedd-4-like ubiquitin-protein ligase atrophin-1 interacting protein 2 Atrophin-1-interacting protein 2 EC 6.3.2 EC 6.3.2.- NEDD4-like E3 ubiquitin-protein ligase WWP2 WW domain containing E3 ubiquitin protein ligase 2 WW domain-containing protein 2 WWp2-like; atrophin-1 interacting protein 2; Atrophin-1-interacting protein 2; HECT-type E3 ubiquitin transferase WWP2; NEDD4-like E3 ubiquitin-protein ligase WWP2; WW domain-containing protein 2
Gene Aliases: AIP2; WWP2; WWp2-like
UniProt ID: (Human) F5H213
Entrez Gene ID: (Human) 11060
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.