Invitrogen
Manufacturer Code:PA528709
Catalog # PIPA528709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1 and 81 of Human WWOX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D16S432E FOR FRA16D HHCMA56 PRO0128 SDR41C1 WOX1 WW domain-containing oxidoreductase WW domain-containing protein WWOX fragile site FRA16D oxidoreductase short chain dehydrogenase/reductase family 41C member 1; Fragile site FRA16D oxidoreductase; Short chain dehydrogenase/reductase family 41C member 1; short chain dehydrogenase/reductase family 41C, member 1; WW domain-containing oxidoreductase; WW domain-containing protein WWOX
Gene Aliases: D16S432E; EIEE28; FOR; FRA16D; HHCMA56; PRO0128; SCAR12; SDR41C1; WOX1; WWOX
UniProt ID: (Human) Q9NZC7
Entrez Gene ID: (Human) 51741
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.