R&D Systems
Manufacturer Code:AF6399SP
Catalog # AF6399SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human WWOX Asn36-Val126 Accession # Q9NZC7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: D16S432E EC 1.1.1 EC 1.1.1.- FOR FORHHCMA56 FRA16D Fragile site FRA16D oxidoreductase SDR41C1 short chain dehydrogenase/reductase family 41C member 1 WOX1 WOX1PRO0128 WW domain containing oxidoreductase WW domain-containing oxidoreductase WW domain-containing protein WWOX; Fragile site FRA16D oxidoreductase; Short chain dehydrogenase/reductase family 41C member 1; short chain dehydrogenase/reductase family 41C, member 1; WW domain-containing oxidoreductase; WW domain-containing protein WWOX
Gene Aliases: D16S432E; EIEE28; FOR; FRA16D; HHCMA56; PRO0128; SCAR12; SDR41C1; WOX1; WWOX
UniProt ID: (Human) Q9NZC7
Entrez Gene ID: (Human) 51741
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.