Invitrogen
Manufacturer Code:PA571284
Catalog # PIPA571284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human WWC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BH-3-only member B; BH3-only member B protein; Protein WWC2; WW domain-containing protein 2; WW, C2 and coiled-coil domain containing 2
Gene Aliases: 5330431C14Rik; AU022508; BOMB; C85457; D8Ertd594e; MNCb-4173; RGD1559427; WWC2
UniProt ID: (Human) Q6AWC2, (Mouse) Q6NXJ0
Entrez Gene ID: (Horse) 100058969, (Dog) 482920, (Pig) 100621011, (Human) 80014, (Rat) 498630, (Mouse) 52357, (Guinea pig) 100719966
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.