Invitrogen
Manufacturer Code:PA5116880
Catalog # PIPA5116880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human WWC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5330431C14Rik AU022508 BH-3-only member B BH3-only member B protein BOMB C85457 D8Ertd594e MNCb-4173 Protein WWC2 WW and C2 domain containing 2 WW domain-containing protein 2 WW C2 and coiled-coil domain containing 2 Wwc2; BH-3-only member B; BH3-only member B protein; Protein WWC2; WW domain-containing protein 2; WW, C2 and coiled-coil domain containing 2
Gene Aliases: 5330431C14Rik; AU022508; BOMB; C85457; D8Ertd594e; MNCb-4173; RGD1559427; WWC2
UniProt ID: (Human) Q6AWC2, (Mouse) Q6NXJ0
Entrez Gene ID: (Human) 80014, (Rat) 498630, (Mouse) 52357
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.