Invitrogen
Manufacturer Code:PA558212
Catalog # PIPA558212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WWC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HBeAg-binding protein 3; KIBRA; Kidney and brain protein; Protein KIBRA; protein phosphatase 1, regulatory subunit 168; protein WWC1; WW and C2 domain containing 1 FLJ10865 FLJ23369 HBEBP3 HBEBP36 KIAA0869 KIBRA; WW domain-containing protein 1; WW, C2 and coiled-coil domain containing 1
Gene Aliases: HBEBP3; HBEBP36; KIAA0869; KIBRA; MEMRYQTL; PPP1R168; WWC1
UniProt ID: (Human) Q8IX03
Entrez Gene ID: (Human) 23286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.