R&D Systems
Manufacturer Code:MAB7374SP
Catalog # MAB7374SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 729419 |
Immunogen | <em>E. coli</em>-derived recombinant human WTX Met421-Leu573 Accession # Q5JTC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: adenomatous polyposis coli membrane recruitment 1; Amer1; APC membrane recruitment protein 1; family with sequence similarity 123B; Protein FAM123B; RP11-403E24.2; Wilms tumor gene on the X chromosome protein; Wilms tumor on the X
Gene Aliases: AMER1; FAM123B; OSCS; WTX
UniProt ID: (Human) Q5JTC6
Entrez Gene ID: (Human) 139285
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.