Proteintech
Manufacturer Code:554321AP150UL
Catalog # 50-173-6155
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenomatous polyposis coli membrane recruitment 1; Amer1; AMER1 FAM123B Protein FAM123B WTX; APC membrane recruitment protein 1; family with sequence similarity 123, member B; family with sequence similarity 123B; Protein FAM123B; RP11-403E24.2; Wilms tumor gene on the X chromosome protein; Wilms tumor on the X
Gene Aliases: 2810002O09Rik; AMER1; AW492303; FAM123B; OSCS; WTX
UniProt ID: (Human) Q5JTC6, (Mouse) Q7TS75
Entrez Gene ID: (Human) 139285, (Mouse) 72345
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.